Learn More
Ahlstrom™ Filter Papers Grade – 320 PROMO
Manufacturer:  AHLSTROM FILTRATION INC 32081515PM
 View more versions of this product
Catalog No. 0979054GPM
Add to cart
Specifications
Specifications
320
15cm
High
15cm
Electrophoresis of large molecules and as blotter to complete the blotting sandwich in gel transfer
2.45mm