Learn More
Ahlstrom™ Filter Papers Grade – 320 PROMO
Manufacturer:  AHLSTROM FILTRATION INC 32082020PM
 View more versions of this product
Catalog No. 0979054KPM
Add to cart
Specifications
Specifications
320
20cm
High
20cm
Electrophoresis of large molecules and as blotter to complete the blotting sandwich in gel transfer
2.45mm