Learn More
Ahlstrom™ Filter Papers – Grade 601
Manufacturer:  Ahlstrom™ 60101100
 View more versions of this product
Catalog No. 09-790-55E
Add to cart
Specifications
Specifications
11cm
Clarifying liquids, rapid filtration of coarse precipitates: lead sulfate, lead chromate, calcium oxalate/carbonate, aluminum phosphate
Medium
601
2.5μm