Learn More
Ahlstrom™ Filter Papers – Grade 601
Manufacturer:  Ahlstrom™ 60100425
 View more versions of this product
Catalog No. 09-800-55A
Add to cart
Specifications
Specifications
4.25cm
Clarifying liquids, rapid filtration of coarse precipitates: lead sulfate, lead chromate, calcium oxalate/carbonate
Medium
601
2.5μm