Learn More
Ahlstrom-Munksjo Standard Non Wet-Strengthened Qualitative Filter Paper, Grade 610
Slow flowing filter paper designated to retain fine particles encountered in qualitative analysis. Widely used for water analysis, biological products and fine precipitates such as cold-precipitate barium sulfate. It is also used in Buchner funnels and to clarify cloudy suspensions.
Supplier:  Ahlstrom-Munksjo 61000900
 View more versions of this product
Catalog No. 09-790-59C
Explore available promotions
Add to Cart
Description
Description
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Disc
Slow
610
1.5 μm
Water analysis, biological products and fine precipitates; also used in Buchner funnels and to clarify cloudy suspensions
9 cm
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.