Learn More
Ahlstrom™ Filter Papers – Grade 615
Manufacturer:  Ahlstrom™ 61500700
 View more versions of this product
Catalog No. 09-790-63
Add to cart
Specifications
Specifications
7cm
Filtering aluminum, ferric, magnesium hydroxides. For construction filtration work, filtration of insoluble iron and ore.
Fast
615
25μm