Learn More
Ahlstrom Filter Papers™ – Grade 617 PROMO
Supplier: Ahlstrom 61701500PM
Specifications
35 μm | |
Rapid filtration of coarse particles and gelatinous precipitates such as in oils and resin solutions; construction product testing | |
15 cm |
Fast | |
617 |