Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ahlstrom Filter Papers™ – Grade 617 PROMO

Catalog No. p-7114623 Shop All Ahlstrom Products
Click to view available options
Diameter (Metric):
90 mm
110 mm
125 mm
150 mm
250 mm
330 mm

Specifications

Particle Retention 35 μm
Flow Rate Fast
For Use With (Application) Rapid filtration of coarse particles and gelatinous precipitates such as in oils and resin solutions; construction product testing
Grade 617
Diameter (Metric) 90 mm
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.