Learn More
Ahlstrom Filter Papers -- Grade 631 PROMO
Manufacturer:  AHLSTROM FILTRATION INC 63101250PM
 View more versions of this product
Catalog No. 0979067EPM
Add to cart
Specifications
Specifications
12.5cm
Filtration of coarse particles and gelatinous precipitates; air monitoring and gas filtration when high flow rate is needed
Medium
631
10μm