Learn More
Ahlstrom™ Filter Papers -- Grade 631
Manufacturer:  Ahlstrom™ 63100700
 View more versions of this product
Catalog No. 09-790-67B
Add to cart
Specifications
Specifications
7cm
Filtration of coarse particles and gelatinous precipitates; air monitoring and gas filtration when high flow rate is needed
Medium
631
10μm