Learn More
Ahlstrom Filter Papers -- Grade 642 PROMO
Manufacturer:  AHLSTROM FILTRATION INC 64201500PM
 View more versions of this product
Catalog No. 0979070FPM
Add to cart
Specifications
Specifications
15cm
General lab applications; agricultural and environmental labs for plant growth trials, soil testing, and air analysis for monitoring contaminants
Slow
642
2μm