Learn More
Ahlstrom™ Glass Microfiber Filter Papers - Grade 111
For general purpose use and atmospheric and water pollution monitoring
Manufacturer:  Ahlstrom™ 11100900
 View more versions of this product
Catalog No. 09-790-33F
Add to cart
Specifications
Specifications
3.5 in.
Circle
1.2μm
500°C
0.28mm
General purpose use and atmospheric/water pollution monitoring
5 cfm/ft2
111
932°F
0.01 in.
9cm
Borosilicate Glass