Learn More
Ahlstrom™ Glass Microfiber Filter Papers - Grade 121 PROMO
For pre-filtration of submicron membranes, clarification of liquid suspensions and LSC techniques
Supplier: Ahlstrom 12100250PM
Specifications
Disc | |
1 μm | |
121 | |
500°C | |
2.5 cm |
Borosilicate Glass | |
Prefiltration of submicron membranes, clarification of liquid suspensions and LSC techniques | |
932°F | |
0.98 in. |