Learn More
Ahlstrom™ Glass Microfiber Filter Papers - Grade 161 PROMO
Ahlstrom Grade 161 is a binder-free microfiber glass filter that offers finer particle retention and higher flow rate than traditional analytical papers.
Supplier:  Ahlstrom 16101100PM
 View more versions of this product
Catalog No. 0979046MPM
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
Description
Description
  • Suitable for Total Solids Testing (TSS) as described in STM 2540D 7
  • Thermal resistance up to 500 degrees Celsius
Specifications
Specifications
Disc
1.1 μm
Fine porosity and high flow for TSS, cell harvesting, and general laboratory applications
932°F
0.012 in.
4.33 in.
Borosilicate Glass
3.6 cfm/ft2
161
500°C
0.32 mm
11 cm
Product Suggestions
Product Suggestions