Learn More
Ahlstrom-Munksjo Quantitative Filter Papers (Ashless), Grade 55
Fast filtering grade for gravimetric analysis of coarse particles and gelatinous precipitates. Also used in air and foodstuffs analysis and for chloride and phosphorous in cement, coal and coke.
Manufacturer:  Ahlstrom-Munksjo 05501500
Catalog No. 22-286641
Add to cart
Description
Description
Specifications
Specifications
15 cm
Analysis of coarse and gelatinous precipitates
55
31 Herzberg sec/100 mL
Circle
15μm