Learn More
Abnova™ AHNAK 293T Cell Transient Overexpression Lysate (Denatured)
Human overexpression lysate
Manufacturer:  Abnova™ H00079026T02
Catalog No. 89-932-906
Add to cart
Description
Description
Specifications
Specifications
293T
NM_024060
pCMV-AHNAK full-length
100 μL
Human
AHNAK 293T Cell Transient Overexpression Lysate(Denatured)
Theoretical M.W. 16.1kDa
Transient overexpression cell lysate was tested with anti-AHNAK antibody (Mfr. No. H00079026-B01P) by Western Blot.
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue)
Western Blot