Learn More
Abnova™ AHNAK2 293T Cell Transient Overexpression Lysate (Denatured) (T03)
Human overexpression lysate
Manufacturer:  Abnova™ H00113146T03
Catalog No. 89-932-908
Add to cart
Description
Description
Specifications
Specifications
293T
BC049216
pCMV-AHNAK2 full-length
100 μL
Human
AHNAK2 293T Cell Transient Overexpression Lysate(Denatured)
Theoretical M.W. 52.7kDa
Transient overexpression cell lysate was tested with anti-AHNAK2 antibody (Mfr. No. H00113146-B01P) by Western Blot.
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue)
Western Blot