Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ AHNAK2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA552061
Description
Immunogen sequence: APRAKLDSAQ LEGDLSLADK DVTAKDSKFK MPKFKMPSFG VSAPGKSIEA SVHVSAPKVE ADVSLPSMQG DLKTTDLSIQ PHSADLTVQA RQVDMKLLEG HVPEEAGLKG HLPKVQMPSF KMPKVDLKG Highest antigen sequence identity to the following orthologs: Mouse - 67%, Rat - 63%.
The specific function of AHNAK2 is not yet known.
Specifications
AHNAK2 | |
Polyclonal | |
Unconjugated | |
AHNAK2 | |
AHNAK nucleoprotein 2; AHNAK2; AI450948; C14orf78; Gm1185; Gm72; KIAA2019; Protein AHNAK2; RGD1309696 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
113146 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Immunocytochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8IVF2 | |
AHNAK2 | |
Recombinant protein corresponding to Human AHNAK2. Recombinant protein control fragment (Product #RP-89107). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction