Learn More
AHR Sheep anti-Human, Polyclonal, R&D Systems™
Sheep Polyclonal Antibody
Manufacturer:  R&D Systems AF6185SP
Catalog No. AF6185SP
Add to cart
This item is not returnable. View return policy
Description
Description
AHR Polyclonal antibody AF6185SP specifically detects AHR in Human samples. It is validated for Flow Cytometry (Intracellular Staining), Immunocytochemistry, Simple Western, Western Blotting
Specifications
Specifications
AHR
LYOPH
Western Blot 1 μg/mL, Simple Western 10 μg/mL, Intracellular Staining by Flow Cytometry 2.5 μg/10^6 cells, Immunocytochemistry 5-15 μg/mL, CyTOF-ready, Knockout Validated
P35869
Ah receptor, AhR, AH-receptor, aryl hydrocarbon receptor, BHLHE76, bHLHe76aromatic hydrocarbon receptor, Class E basic helix-loop-helix protein 76
Sheep
IgG
25 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects human AHR in Western blots.
Human
Western Blot, Flow Cytometry, Immunocytochemistry, CyTOF, Immunoassay
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
AHR
E. coli-derived recombinant human AHR Asn704-Leu848 Accession # P35869
Antigen Affinity-purified
RUO
Primary
196
Reconstitute at 0.2 mg/mL in sterile PBS.