Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
AHSA1 Antibody (25F2.D9), Alexa Fluor™ 532, Novus Biologicals™
Rat Monoclonal Antibody
Supplier: Novus Biologicals NBP212883AF532
Description
AHSA1 Monoclonal specifically detects AHSA1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
AHSA1 | |
Monoclonal | |
Alexa Fluor 532 | |
50 mM sodium borate with 0.05% sodium azide | |
AHSA1 | |
Recombinant Full Length Mouse Aha1 Protein | |
0.1 mL | |
10598.0 | |
Human, Mouse, Rat | |
IgG2a κ |
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
25F2.D9 | |
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin | |
activator of 90 kDa heat shock protein ATPase homolog 1, AHA1, activator of heat shock 90kDa protein ATPase homolog 1 (yeast), C14orf3, chromosome 14 open reading frame 3, p38AHA1 | |
Rat | |
Protein G purified | |
Primary | |
Detects approx 38 kDa. Can run up to 45 kDa on SDS-PAGE. | |
Store at 4°C in the dark. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction