Learn More
AHSA1 Rat anti-Human, Mouse, Rat, Alexa Fluor™ 594, Clone: 25F2.D10, Novus Biologicals™
Rat Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP212884AF594
Catalog No. NP212884A94
Add to cart
Description
Description
AHSA1 Monoclonal antibody specifically detects AHSA1 in Human,Mouse,Rat samples. It is validated for Western Blot,ELISA,Immunohistochemistry,Immunocytochemistry,Immunofluorescence,Immunohistochemistry (Paraffin).
Specifications
Specifications
AHSA1
25F2.D10
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
activator of 90 kDa heat shock protein ATPase homolog 1, AHA1, activator of heat shock 90kDa protein ATPase homolog 1 (yeast), C14orf3, chromosome 14 open reading frame 3, p38AHA1
Rat
IgG2a Kappa
Store at 4°C in the dark.
Monoclonal
Detects approx 38kDa. Can run up to 45kDa on SDS Page.
ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Alexa Fluor™ 594
50 mM sodium borate with 0.05% sodium azide
AHSA1
Mouse Aha1
0.1 mL
Primary
10598
Human, Mouse, Rat