Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ AI-3 Available on GSA/VA Contract for Federal Government customers only.

ARE activator; induces Nrf2-ARE-dependent transcription

$180.00 - $749.00

Specifications

Product Type AI-3
Inhibitors AI-3
Molecular Weight 292.8
Purity >98
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
51-451-0 Available on GSA/VA Contract for Federal Government customers only.
View Documents
Tocris Bioscience™
5145/10
10mg
Each for $180.00
Add to cart
 
51-455-0
View Documents
Tocris Bioscience™
5145/50
50mg
Each for $749.00
Add to cart
 
Description

Description

Antioxidant response element (ARE) activator; disrupts Nrf2/Keap1 and Keap1/Cul3 interactions and stabilizes Nrf2. Activates ARE in an Nrf2 and PI 3-kinase dependent manner. Induces cytoprotective gene expression in vitro and in vivo.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Specifications

AI-3
AI-3
292.8
>98
SDS

For Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit