Tocris™ AI-3

ARE activator; induces Nrf2-ARE-dependent transcription

Manufacturer: Tocris™ 5145/10

 View more versions of this product

Catalog No. 51-451-0

  Check Availability
Add to Cart

Disclaimers:

For Research Use Only


    Specifications

    Inhibitors AI-3
    Purity >98
    Molecular Weight 292.8
    Quantity 10mg
    Product Type AI-3

    Antioxidant response element (ARE) activator; disrupts Nrf2/Keap1 and Keap1/Cul3 interactions and stabilizes Nrf2. Activates ARE in an Nrf2 and PI 3-kinase dependent manner. Induces cytoprotective gene expression in vitro and in vivo.

    • Chemical Name: 1-Chloro-6,7-dihydro-6,6-dimethyl-3-(methylsulfonyl)-benzo[c]thiophen-4(5H)-one
    • Formula: C11H13ClO3S2
    • CAS Number: 882288-28-0
    • Solubility: Soluble to 100mM in DMSO and to 50mM in ethanol
    • Purity: >98

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit