Learn More
Akina IncSupplier Diversity Partner AI20 PLGA-PEG-MAL 100mg DFS Item
Manufacturer:  Akina IncSupplier Diversity Partner AI20
Poly(lactide-co-glycolide)-b-poly(ethylene Glycol)-maleimide (20,000:5,000 Da)
Catalog No. 50-489-377