Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AIB-1 Mouse, Unlabeled, Clone: 34, BD

Mouse Monoclonal Antibody

Manufacturer:  BD Biosciences 611104

 View more versions of this product

Catalog No. BDB611104


Add to cart

Description

Description

Signal transduction via nuclear hormone receptors is important for cell growth and differentiation, development, and homeostasis. Nuclear hormone receptors are ligand-activated transcription factors that modulate target gene expression. These ligand/receptor complexes also interact with transcriptional coactivators which enhance ligand-dependent transcription. Various classes of coactivators have been identified, including SRC-1 and its related proteins, such as TIF-2/GRIP-1, RIP140, AIB-1, and TIF-1α and -1β. AIB-1 (Amplified In Breast cancer-1) is also known as pCIP, RAC3, and TRAM-1. It interacts with estrogen receptor (ER) and is overexpressed in breast cancer biopsies and several breast and ovarian cancer cell lines. Similar to SRC-1 and TIF2, AIB-1 contains a basic helix-loop-helix (bHLH) domain followed by a PAS (Per/Arnt/Sim) region, serine and threonine rich regions, and a charged cluster. In addition, AIB-1 contains three copies of the conserved LXXLL motif which is critical to its interaction with the nuclear receptor. Thus, AIB-1 is a coactivator of nuclear receptors that may participate in the development of steroid-dependent cancers.

Specifications

Specifications

AIB-1
34
Unconjugated
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Murine
IgG1
RUO
Store undiluted at -20°C.
Monoclonal
Western Blot
250μg/mL
Amplified In Breast cancer-1; pCIP; RAC3; TRAM-1
Affinity Purified
Human AIB-1 aa. 376-389
50μg
Cell Biology
Primary
Human, Murine, Rat
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only.