Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

APOA1BP Rabbit anti-Human, Mouse, Rat, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA549569

Catalog No. PA549569

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

The product of this gene interacts with apolipoprotein A-I , the major apolipoprotein of high-density lipoproteins . It is secreted into some bodily fluids, and its synthesis and secretion are stimulated in vitro by incubating cells with apoA-I. The human genome contains related pseudogenes.
Specifications

Specifications

APOA1BP
Polyclonal
PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4
Q8NCW5, Q8K4Z3, B0BNM1
NAD(P)H-hydrate epimerase, Apolipoprotein A-I-binding protein, AI-BP, NAD(P)HX epimerase, YjeF N-terminal domain-containing protein 1, YjeF_N1, APOA1BP
Rabbit
IgG
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
128240, 246703, 295229
Immunocytochemistry, Immunofluorescence, Western Blot
Unconjugated
NAXE
Liquid
NAXE
synthetic peptide corresponding to133-161aa of human NNRE
Affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit