Learn More
Abnova™ AICDA 293T Cell Transient Overexpression Lysate (Denatured)
Human overexpression lysate
Manufacturer:  Abnova™ H00057379T02
Catalog No. 89-932-925
Add to cart
Description
Description
Specifications
Specifications
293T
NM_020661.1
pCMV-AICDA full-length
100 μL
Human
AICDA 293T Cell Transient Overexpression Lysate(Denatured)
Theoretical M.W. 24kDa
Transient overexpression cell lysate was tested with anti-AICDA antibody (Mfr. No. H00057379-D01P) by Western Blot.
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue)
Western Blot