Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AIDA Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548884

Catalog No. PA548884

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Predicted to react with zebrafish, monkey and mouse based on sequence homology.

Acts as a ventralizing factor during embryogenesis. Inhibits axin-mediated JNK activation by binding axin and disrupting axin homodimerization. This in turn antagonizes a Wnt/beta-catenin-independent dorsalization pathway activated by AXIN/JNK-signaling.
Specifications

Specifications

AIDA
Polyclonal
PBS with 0.09% sodium azide
Q96BJ3
Axin interactor, dorsalization-associated protein, Axin interaction partner and dorsalization antagonist, AIDA, C1orf80
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
64853
Western Blot
Unconjugated
AIDA
Liquid
AIDA
KLH conjugated synthetic peptide between 255-283 amino acids from the C-terminal region of human AIDA.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit