Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
AIF-1/Iba1, Mouse anti-Human, Alexa Fluor 405, Clone: rAIF1/1909, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer: Novus Biologicals NBP275760AF405
Description
AIF-1/Iba1 Monoclonal antibody specifically detects AIF-1/Iba1 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin).Specifications
AIF-1/Iba1 | |
rAIF1/1909 | |
Immunohistochemistry, Immunohistochemistry-Paraffin | |
50mM Sodium Borate with 0.05% Sodium Azide | |
AIF1 | |
AIF-1/Iba1 Antibody (rAIF1/1909) was developed against purified recombinant human AIF1 protein (around aa 1-146) (exact sequence is proprietary) | |
Monoclonal | |
Protein A or G purified | |
RUO | |
Store at 4°C in the dark. | |
AIF1 is a cytoplasmic, calcium-binding protein that is thought to play a role in macrophage activation and function. AIF1, containing two EF domains, is induced by cytokines and Interferons. In an unstimulated state, AIF1 colocalizes with actin, and upon stimulation, translocates to lamellipodia. It is also a marker of human microglia and is expressed by macrophages in injured skeletal muscle. The gene encoding AIF1 resides in the tumor necrosis factor (TNF) cluster of genes, located in the region represented by the human major histocompatibility complex (MHC). |
Immunohistochemistry,Immunohistochemistry (Paraffin) | |
Alexa Fluor 405 | |
Purified,Conjugated | |
AIF-1, allograft inflammatory factor 1, G1, IBA1, IbaI, interferon gamma responsive transcript, Ionized calcium-binding adapter molecule 1, IRT1, IRT-1, Protein G1 | |
Mouse | |
IgG1 | |
Primary | |
0.1 mL | |
Autoimmune Diseases, Cancer, Cell Biology, Cell Cycle and Replication, Immunology, Inflammation, Lipid and Metabolism, Microglia Markers, Neuroscience | |
Human |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title