Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AIF1 Mouse anti-Human, FITC, Clone: 1022-5, Abnova™

Mouse monoclonal antibody raised against human AIF1.

Manufacturer:  Abnova Corporation MAB12585

Catalog No. 89-933-529


Add to cart

Description

Description

This gene is induced by cytokines and interferon. Its protein product is thought to be involved in negative regulation of growth of vascular smooth muscle cells, which contributes to the anti-inflammatory response to vessel wall trauma. Three transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq]

Specifications

Specifications

AIF1
1022-5
Mouse monoclonal antibody raised against human AIF1.
In PBS, pH 7.2 (10mg/mL BSA, 0.09% sodium azide).
AIF-1/IBA1/IRT-1
Mouse
IgG2b
50 ug
Store at 4°C. Do not freeze.
Monoclonal
Human
Immunohistochemistry (PFA fixed)
FITC
Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:50) The optimal working dilution should be determined by the end user.
Liquid
AIF1
Recombinant protein corresponding to human AIF1.
Affinity purification
RUO
Primary
199
SDS
Documents
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit