Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AIG1 Rabbit anti-Human, Polyclonal , Abnova™

AIG1 polyclonal antibody raised against recombinant human AIG1.

Manufacturer:  Abnova Corporation PAB29478

Catalog No. 89-933-530


Add to cart

Description

Description

Sequence: AIEMPSHQTYGGSWKFLTFID
Specifications

Specifications

AIG1
Unconjugated
Immunofluorescence (1-4 ug/mL) Immunohistochemistry (1:20-1:50) The optimal working dilution should be determined by the end user.
Liquid
AIG1
Recombinant protein corresponding to amino acids of human AIG1.
Antigen affinity purification
RUO
Primary
51390
Immunofluorescence, Immunohistochemistry (PFA fixed)
AIG1 polyclonal antibody raised against recombinant human AIG1.
In PBS, pH 7.2 (40% glycerol, 0.02% sodium azide)
AIG-1/DKFZp686F03136/FLJ10485/dJ95L4.1
Rabbit
IgG
100 uL
Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Polyclonal
Human
SDS
Documents
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit