Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
AIM1L Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA548761
Description

Specifications
AIM1L | |
Polyclonal | |
PBS with 0.09% sodium azide | |
Q8N1P7 | |
CRYBG2 | |
KLH conjugated synthetic peptide between 395-423 amino acids from the C-terminal region of human AIM1L. | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
CRYBG2 | |
Absent in melanoma 1-like protein, Beta/gamma crystallin domain-containing protein 2, AIM1L, CRYBG2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
55057 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction