Learn More
Abnova™ AIM2 293T Cell Transient Overexpression Lysate (Denatured) (T04)
Human overexpression lysate
Manufacturer:  Abnova™ H00009447T04
Catalog No. 89-932-929
Add to cart
Description
Description
Specifications
Specifications
293T
NM_004833
pCMV-AIM2 full-length
100 μL
Human
AIM2 293T Cell Transient Overexpression Lysate(Denatured)
Theoretical M.W. 39kDa
Transient overexpression cell lysate was tested with anti-AIM2 antibody (Mfr. No. H00009447-B01P) by Western Blot.
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue)
Western Blot