Learn More
AIM2 Mouse anti-Human, Alexa Fluor™ 350, Clone: 10M2B3, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP227355AF350
Catalog No. NP2273A30
Add to cart
Description
Description
AIM2 Monoclonal antibody specifically detects AIM2 in Human samples. It is validated for Western Blot.
Specifications
Specifications
AIM2
10M2B3
Western Blot
absent in melanoma 2PYHIN4, interferon-inducible protein AIM2
Mouse
IgG2a Kappa
Store at 4°C in the dark.
Monoclonal
Human
Western Blot
Alexa Fluor™ 350
50 mM sodium borate with 0.05% sodium azide
AIM2
This antibody was raised against recombinant human AIM2 protein
0.1 mL
Primary
9447