Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AIM2 Mouse anti-Human, Alexa Fluor™ 750, Clone: 10M2B3, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP227355AF750

Catalog No. NP2273A70


Add to cart

Description

Description

AIM2 Monoclonal antibody specifically detects AIM2 in Human samples. It is validated for Western Blot.
Specifications

Specifications

AIM2
10M2B3
Western Blot
Conjugated, Purified
AIM2
This antibody was raised against recombinant human AIM2 protein
Protein G purified
Store at 4°C in the dark.
Monoclonal
Human
Western Blot
Alexa Fluor™ 750
50 mM sodium borate with 0.05% sodium azide
absent in melanoma 2PYHIN4, interferon-inducible protein AIM2
Mouse
IgG2a Kappa
0.1 mL
Primary
9447.0
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit