Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AIMS™ AIMS™ ATS-3 General Rodent Tattoo System Available on GSA/VA Contract for Federal Government customers only.

Lab animal tattoo identification system

Manufacturer:  AIMS™ ATS3

 View more versions of this product

Catalog No. 14-370-130


Add to cart

Description

Description

Comes complete with everything you need to start tattooing weanling or older mice and rats.

ATS-3 General Rodent Tattoo System (14-370-130)

  • Includes supplies and accessories for tattooing approx. 3000 mice/rats
  • Detailed training guide and instruction DVD provide step-by-step guidance
  • Use numbers, letters, lines, dots, and/or colors to permanently ID any color/strain mouse or rat
  • The most versatile, basic tattoo identification system for laboratory mice and rats
  • Heavy duty custom equipment
  • Humane mouse and rat restraints
  • Tattoo weanling to adult mice/rats
  • Reduce animal stress; quick and safe
  • Foot pedal ON/OFF; preset speeds
  • No anesthesia needed
  • Economical; costs about 5 cents/rodent
  • Unlimited tech support
  • Standard System Container: 20L x 10W x 10 in.D
  • Tattoo Machine Controller: 5L x 5W x 2 in.D (mounted in top tray 15.5L x 7.75W x 2 in.D)
  • Power Supply: 3.25L x 1.75W x 1 in.D (mounted in top tray 15.5L x 7.75W x 2 in.D)
  • Universal power supply
  • Power cord supplied for individual country needs
  • Input 100-220V, 50-60 Hz
LLA-AP Large Lab Animal Upgrade Package (14-370-133)
  • Includes accessories and supplies for large lab animal tattooing including Volume 3 of Laboratory Animal Tattoo Identification, Large Lab Animals

 

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Compliance

UI, CE, globally approved. RoHS compliant.

Specifications

Specifications

Supplies and accessories for tattooing 3000 mice/rats, training guide, instruction DVD, tattoo identification system, standard system container, tattoo machine controller, custom equipment, humane mouse and rat restraints, tattoo weanling, power supply
25.4cm
20 in.
50.8cm
25.4cm
20 x 10 in. (50.8 x 25.4cm)
UI, CE, RoHS
10 in.
Ideal for tattooing 3000 mice or rats
50/60Hz
100/220V
10 in.
100/220V 50/60Hz
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit