Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AIMS™ Tissue Processing Discs

For dissecting and inspecting tissue samples

Manufacturer:  AIMS™ LTPD

 View more versions of this product

Catalog No. 14-371-171


Add to cart

Description

Description

  • Process tissue using clean blue wax surface disc
  • Layout and pin tissue on surface using ball head tissue pins
  • Disc can reused and resurfaced multiple times if desired
  • Thickness: 0.25 in.

Specifications

Specifications

Blue
10cm
Tissue Processing Disc
Environmentaly Friendly Blue Wax
Large
4 in.
Tissue processing
AIMS Procedure/Dissection Boards
Circular
6.35mm (0.25 in.)
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit