Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aiolos/IKZF3 Mouse anti-Human, Alexa Fluor 350, Clone: 914821, R&D Systems™

Mouse MonoclonalAntibody

Manufacturer:  R&D Systems FAB8625U100UG

Catalog No. FAB8625U100


Add to cart
This item is not returnable. View return policy

Description

Description

Recombinant Monoclonal Antibody.
Specifications

Specifications

Aiolos/IKZF3
Monoclonal
0.2 mg/mL
Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells
Q9UKT9
AIO, AIOLOS, IKAROS family zinc finger 3 (Aiolos), Ikaros family zinc finger protein 3, IKZF3, zinc finger protein Aiolos, ZNFN1A3, ZNFN1A3subfamily 1A, 3 (Aiolos)
Mouse
IgG1
100 ug
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Monoclonal
Detects human Aiolos/IKZF3 in direct ELISAs.
Flow Cytometry
914821
Alexa Fluor 350
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
Purified
IKZF3
E. coli-derived recombinant human Aiolos/IKZF3 Met1-Gly54 Accession # Q9UKT9
Protein A or G purified from cell culture supernatant
RUO
Primary
22806
Human
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only (RUO)