Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aiolos/IKZF3 Mouse anti-Human, Janelia Fluor™ 549, Clone: OTI7E11, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP271698JF549

Catalog No. N271698J549


Add to cart

Description

Description

Aiolos/IKZF3 Monoclonal antibody specifically detects Aiolos/IKZF3 in Human samples. It is validated for Immunohistochemistry.
Specifications

Specifications

Aiolos/IKZF3
OTI7E11
Immunohistochemistry
Affinity Purified, Conjugated
IKZF3
Recombinant protein fragment corresponding to amino acids 1-301 of human Aiolos/IKZF3 (NP_036613) produced in E.coli.
Immunogen affinity purified
Chromatin Research
Primary
22806.0
Immunohistochemistry
Janelia Fluor 549
50mM Sodium Borate with 0.05% Sodium Azide
AIO, AIOLOS, IKAROS family zinc finger 3 (Aiolos), Ikaros family zinc finger protein 3, zinc finger protein Aiolos, ZNFN1A3subfamily 1A, 3 (Aiolos)
Mouse
IgG2b
0.1 mL
Store at 4°C in the dark.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit