Learn More
Environmental Express™ Air Filter Cellulose Support Pads
Ideal for air particle monitoring and environmental testing
Manufacturer:  Environmental Express™ PG37100
Catalog No. 09-732-48
Add to cart
Description
Description
  • Laser cutting ensures a clean, fiber-free filter edge
  • Excellent temperature and chemical resistance
Specifications
Specifications
Gridded
Air Particle Monitoring and Environmental Testing
Cellulose
37 mm
Support Pad