Learn More
Thermo Scientific™ Air Filter Kits
Protect your recirculating chillers from dusty environments with these Air Filter Kits.
Manufacturer:  Thermo Scientific™ 598000000014
 View more versions of this product
Catalog No. 13-100-768
Add to cart
Description
Description
Protect your recirculating chillers from dusty environments with Thermo Scientific™ Air Filter Kits.
For use with Thermo Scientific recirculating chillers.
Specifications
Specifications
Air and Fluid Filter
Air and fluid filter kit (100 micron)
ThermoFlex™ 7500 or 1000 Recirculating Chiller