Learn More
Thermo Scientific™ Air Filter Kits
GSA_VA
Protect your recirculating chillers from dusty environments with these Air Filter Kits.
$133.00 - $486.00
Products 8
Catalog Number Mfr. No. For Use With (Equipment) Description Price Quantity & Availability  
Catalog Number Mfr. No. For Use With (Equipment) Description Price Quantity & Availability  
13-265-247
GSA_VA
View Documents
Thermo Scientific™
598000000005
ThermoFlex™ 900 or 1400 Recirculating Chiller Air and fluid filter kit (35 micron)
Each for $214.50
Only null left
Add to Cart
13-265-248
GSA_VA
View Documents
Thermo Scientific™
598000000006
ThermoFlex™ 2500 Recirculating Chiller Air and fluid filter kit (100 micron)
Each for $242.50
Only null left
Add to Cart
13-265-249
Accessories View Documents
Thermo Scientific™
598000000007
ThermoFlex™ 2500 Recirculating Chiller Air and fluid filter kit (35 micron)
Each for $242.50
Only null left
Add to Cart
13-265-250
View Documents
Thermo Scientific™
598000000008
ThermoFlex™ 3500 or 5000 Recirculating Chiller Air and fluid filter kit (100 micron)
Each for $362.50
Only null left
Add to Cart
13-265-251
GSA_VA
View Documents
Thermo Scientific™
598000000009
ThermoFlex™ 3500 or 5000 Recirculating Chiller Air and fluid filter kit (35 micron)
Each for $388.50
Only null left
Add to Cart
13-100-768
View Documents
Thermo Scientific™
598000000014
ThermoFlex™ 7500 or 1000 Recirculating Chiller Air and fluid filter kit (100 micron)
Each for $470.00
Only null left
Add to Cart
13-265-252
View Documents
Thermo Scientific™
598000000013
ThermoFlex™ 7500 or 1000 Recirculating Chiller Air and fluid filter kit (35 micron)
Each for $486.00
Only null left
Add to Cart
13-100-782
View Documents
Thermo Scientific™
096270
Polar Accel 250 & 500 Cooling/Heating Recirculating Chillers Air filter
Each for $133.00
Only null left
Add to Cart
Description
Description
Protect your recirculating chillers from dusty environments with Thermo Scientific™ Air Filter Kits.
For use with Thermo Scientific Neslab HX, CFT or M series recirculating chillers.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.