Learn More
LaMotte™ Air Quality Testing Outfit Products for Science Education
Six test modules analyze key atmospheric pollutants.
Includes:
  • Labware and reagents for 25 tests per test module
  • Portable air sampling pump in a carrying case
  • Three types of impingers
  • Detailed instruction manual
Products 1
Catalog Number Mfr. No. Description Price Qty Availability  
Catalog Number Mfr. No. Description Price Qty Availability  
S66513 Products for Science Education
View Documents
LaMotte™
5960P
N/A Complete outfit This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description
Description
  • Measures ammonia, carbon monoxide, chlorine, hydrogen sulfide, nitrogen dioxide, and sulfur dioxide
  • Diaphragm vacuum pump draws air sample through the absorbing reagent in the impinger at a constant rate between 0 and 2L per minute
  • Integral flowmeter regulates airflow rate to control the amount of air sampled
  • Rugged carrying case houses all necessary components
  • Operates up to 48 hours on four C batteries
Order Info
Grades 6+