Learn More
Air Systems™ Auto-Air™ Breather Box™ Air Filtration System Accessory, Calibration Kit
Kits for Air Systems Carbon Monoxide Monitor
Manufacturer:  Air Systems InternationalSupplier Diversity Partner BBK20103
 View more versions of this product
Catalog No. 19-806-331
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Carbon monoxide monitor
Use to monitor calibration on a monthly basis
Large
Large Calibration Kit
2 pressurized 103l disposable cylinders -one cylinder contains zero air, the other contains 20ppm of co air, flow regulator, pressure gauge, vinyl tubing in hard carry case
missing translation for 'provideContentCorrection'
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
missing translation for 'productTitle'