Learn More
Air Systems™ Auto-Air™ Breather Box™ Air Filtration System Accessory, Particulate Filters
For use with Auto-Air Breather Box
Manufacturer:  Air Systems™ BB50C
 View more versions of this product
Catalog No. 19-806-319
Add to cart
Specifications
Specifications
C-style filter element and o-ring
Safety
Removes oil and particulate to 99.9998% at 0.01μ
Bb50-co series breather box™
Oil Coalescing Filter