Learn More
Air Systems™ Auto-Air™ / Breather Box™ Air Filtration System Accessory, Remote Alarm
Designed to duplicate local alarms found on primary equipment to signal workers in a remote location of an alarm condition with their system
Manufacturer:  Air Systems™ 50ALMSTAA
Catalog No. 19-806-294
Add to cart
Includes: 50 foot cable, 9-12 VDC
Description
Description
  • Dual audible alarm
  • Mounted visual strobe
Specifications
Specifications
Safety
9 to 12VDC
Alarm
50 foot cable, 9-12 VDC
Auto-Air, Breather Box