Learn More
Air Systems Auto-Air Intrisically Safe Portable Breathing Air Filtration Boxes
Grade-D filtration system designed to provide breathing air for specific number of workers
Manufacturer:  Air Systems InternationalSupplier Diversity Partner BB100COIS
 View more versions of this product
Catalog No. 19-806-315
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Air Filtration System
4 couplings, 2 outlets, filter change indicator, safety relief valve, solid aluminum block manifold, visual flow meter, electrochemical sensor
115V
123cfm
Filter the incoming air from compressor to supply respirator with grade d quality air and monitors the carbon monoxide level present in the air, provide breathing air for a specific number of workers
150psi
OSHA 1910.134, CSA
99.99% at 0.01μ