Air Systems Cascade Air Cylinders
For use with 19-806-340 Bottle Fill Assembly
Manufacturer: Air Systems InternationalSupplier Diversity Partner AC300
 View more versions of this product
Catalog No. 19-121-173
  Check Availability
Add to Cart
    Specifications
    Product Type Air Cylinder
    For Use With (Application) For storage of breathing air and working in hazardous locations or emergency response
    Material Steel
    Pressure 2400psi
    Capacity 300 cu. ft., 8.49cm3
    Capacity (Metric) 8.49cm3
    Capacity (English) 300 cu. ft.
  • Description & Specifications
Related Products
Product Content Feedback