Learn More
Air Systems™ SCBA Accessory, Cascade Bottle Fill Assembly
For storage of high pressure breathing air
Manufacturer:  Air Systems™ CBF4347NB
Catalog No. 19-806-340
Add to cart
Description
Description
Fill whip cylinders and air cylinders sold separately.
Specifications
Specifications
Safety
5000psi
Bottle Fill Assembly
5 ft. fill whip with gauge (high pressure), a hand-tight nut (universal), an on/off button and a bleed valve
Self Contained Breathing Apparatus
For refilling 2216 or 4500psi scba air cylinders