Learn More
Air Systems Cascade Bottle Fill Whip Cylinders
For use with Bottle Fill Assembly
Manufacturer:  Air Systems InternationalSupplier Diversity Partner HPFW5346
 View more versions of this product
Catalog No. 19-821-856
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Install on cga-346 and cga-347 valves
For filling SCBA cylinders
3000psi
60 in.
Fill Whip Cylinder
Gauge, universal hand tight nut, on or off and bleed valve
152.4cm